Cedertræsolie
Glass bottle, 500 mL
Catalog # TCIAC0626-500ML
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: Cedar wood oil, Cedar Oil, Cedar Leaf Oil
- CAS Number:8000-27-9
- MDL Number:MFCD00132766
- Density:0,98 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
Specifications
Specification Test Results
- Appearance:White to Yellow to Orange clear liquid
- Acid Value:max. 20.0
Specifications
- Pakning:Glass bottle
- Pakn:500 mL



