Dextran, M.W. ~ 40000 (Dextran 40)
Glass bottle, 25 g
Katalog # TCIAD1448-25G
Leverandør: TCI
Gå til:
Product Details & Documents
- CAS nummer:9004-54-0
- Formel:(C₆H₁₀O₅)n
- MDL nummer:MFCD00130935
- Merck Index:14,02948
- EINECS:232-677-5
Specifikationer
Specifikationstestresultater
- Appearance:White powder to crystal
- Specific rotation [a]20/D:+193 to +201 deg (C=6, H2O) (calcd.on dried substance)
- Solubility in hot Water:almost transparency
- Drying loss:max. 5.0 %
- limiting viscosity:0.16 to 0.19 cp (after drying)
Specifikationer
- Pakning:Glass bottle
- Pakn:25 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



