Dextran, M.W. ~ 70000 (Dextran 70)
Glass bottle, 25 g
Leverandør: TCI
Gå til:
Product Details & Documents
- CAS nummer:9004-54-0
- Formel:(C₆H₁₀O₅)n
- MDL nummer:MFCD00130935
- Merck Index:14,02948
- EINECS:232-677-5
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White to Almost white powder to crystaline
- Specific rotation [a]20/D:+170 to +200 deg (C=6, H2O)
- pH:5.0 to 7.0 (60g/L, 25deg-C)
- limiting viscosity:0.21 to 0.26 cp
Specifikationer
- Pakning:Glass bottle
- Pakn:25 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



