D(-)-Fructose ≥99.0% (by HPLC)
Glass bottle, 25 g
Katalog # TCIAF0060-25G
Leverandør: TCI
Gå til:
Product Details & Documents
Synonymer: D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
- CAS nummer:57-48-7
- Formel:C₆H₁₂O₆
- MDL nummer:MFCD00148910
- Molecular Weight:180.16 g/mol
- Merck Index:14,04273
- Kogende Pt:552 °C (1013 hPa)
- Smeltende Pt:100…110 °C
- Tæthed:1,59 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-333-3
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White powder to crystal
- Purity (HPLC) (CAD):min. 99.0 area%
- Specific rotation [a]20/D:-90.0 to -94.0 deg (C=4, H2O) (after drying)
- Solubility in Water:almost transparency
Specifikationer
- Pakning:Glass bottle
- Pakn:25 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



