D(-)-Fructose ≥99.0% (by HPLC)
Glass bottle, 500 g
Katalog # TCIAF0060-500G
Leverandør: TCI
Gå til:
Product Details & Documents
Synonymer: D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
- CAS nummer:57-48-7
- Formel:C₆H₁₂O₆
- MDL nummer:MFCD00148910
- Molecular Weight:180.16 g/mol
- Merck Index:14,04273
- Kogende Pt:552 °C (1013 hPa)
- Smeltende Pt:100…110 °C
- Tæthed:1,59 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-333-3
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White powder to crystal
- Purity (HPLC) (CAD):min. 99.0 area%
- Specific rotation [a]20/D:-90.0 to -94.0 deg (C=4, H2O) (after drying)
- Solubility in Water:almost transparency
Specifikationer
- Pakning:Glass bottle
- Pakn:500 g



