Prednison ≥96.0% (by HPLC)
Glass bottle, 25 g
Katalog # TCIAP1276-25G
Leverandør: TCI
Gå til:
Product Details & Documents
- CAS nummer:53-03-2
- Formel:C₂₁H₂₆O₅
- MDL nummer:MFCD00003608
- Molecular Weight:358.43 g/mol
- Merck Index:14,07722
- Smeltende Pt:230…232 °C
- EINECS:200-160-3
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 96.0 area%
- Specific rotation [a]20/D:+165.0 to +175.0 deg (C=0.5, Dioxane)
- Solubility in N,N-DMF:very faint turbidity
Specifikationer
- Pakning:Glass bottle
- Pakn:25 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



