Prednison ≥96.0% (by HPLC)
Glass bottle, 5 g
Katalog # TCIAP1276-5G
Leverandør: TCI
Gå til:
Product Details & Documents
- CAS nummer:53-03-2
- Formel:C₂₁H₂₆O₅
- MDL nummer:MFCD00003608
- Molecular Weight:358.43 g/mol
- Merck Index:14,07722
- Smeltende Pt:230…232 °C
- EINECS:200-160-3
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 96.0 area%
- Specific rotation [a]20/D:+165.0 to +175.0 deg (C=0.5, Dioxane)
- Solubility in N,N-DMF:very faint turbidity
Specifikationer
- Pakning:Glass bottle
- Pakn:5 g



