Beer bottles
200 ml, flip-top, 56 mm, 176 mm, round, klar
Katalog # 215-2953
Leverandør: BERLIN PACKAGING RIXIUS GMBH
Gå til:
Product Details & Documents
Glass, clear for clamp closure.
Ideal for transport and storage of beer samples.
Specifikationer
- Udv.Ø:56 mm
- Gevind:flip-top
- Model:round
- Kapacitet:200 ml
- Farve:klar
- Højde:176 mm