Thiomorpholin ≥98.0% (by GC)
Glass bottle, 5 mL
Katalog # TCIAT1007-5ML
Leverandør: TCI
Gå til:
Product Details & Documents
- CAS nummer:123-90-0
- Formel:C₄H₉NS
- MDL nummer:MFCD00005974
- Molecular Weight:103.19 g/mol
- Merck Index:14,09300
- Kogende Pt:169°C
- Flash Pt:63°C
- EINECS:204-660-2
- FN:3267
- ADR:8,III
Specifikationer
Specifikationstestresultater
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Specific gravity (20/20):1.0890 to 1.0930
- Refractive index n20/D:1.5370 to 1.5400
Specifikationer
- Pakning:Glass bottle
- Pakn:5 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



