Polymyxin Bsulfat
Glass bottle, 5 g
Katalog # TCIAP1923-5G
Leverandør: TCI
Gå til:
Product Details & Documents
- CAS nummer:1405-20-5
- Formel:C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
- MDL nummer:MFCD00131991
- Molecular Weight:1385.61 g/mol
- Merck Index:14,07573
- Opbevaringstemperatur:Køleskab
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White to Orange to Green powder to crystal
- Total Nitrogen:15.0 to 16.0 % (calcd.on anh.substance)
- Specific rotation [a]20/D:-76.0 to -82.0 deg (C=2, H2O) (calcd.on anh.substance)
- Water:max. 10.0 %
- Phenylalanine:9.0 to 12.0 %
Specifikationer
- Pakning:Glass bottle
- Pakn:5 g



