Span® 80
Glass bottle, 25 g
Catalog # TCIAS0060-25G
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: Arlacel 80, Sorbitan monooleate
- CAS Number:1338-43-8
- Formula:C₂₄H₄₄O₆
- MDL Number:MFCD00080948
- Molecular Weight:428.61 g/mol
- Density:1 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:215-665-4
Specifications
Specification Test Results
- Appearance:Yellow to Amber to Dark purple clear liquid
- Saponification Value:142.0 to 189.0
- Acid Value:max. 10.0
Specifications
- Pakning:Glass bottle
- Pakn:25 g



