Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Bromphenol blå natriumsalt

Glass bottle, 5 g
Catalog # TCIAS0043-5G
Supplier:   TCI
Bromphenol blå natriumsalt
Catalog # TCIAS0043-5G
Supplier:  TCI
CAS Number:  34725-61-6
Bromphenol blå natriumsalt
Bromphenol blå natriumsalt

Glass bottle, 5 g

CAS Number:  34725-61-6
Price
1.060,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt
  • CAS Number:34725-61-6
  • Formula:C₁₉H₉Br₄NaO₅S
  • MDL Number:MFCD00013793
  • Molecular Weight:691.95 g/mol
  • Storage Temperature:Stue temperatur
  • EINECS:252-170-2
Specifications

Specification Test Results

  • Appearance:
    Dark green to Dark blue to Black powder to crystal
  • Absorbance:
    min. 0.480 (5 mg/L, pH 5.0, 590.0 to 594.0 nm)
  • Solubility in Water:
    almost transparency
  • Drying loss:
    max. 7.0 %
  • Ignition residue (Sulfate):
    10.0 to 10.8 % (after drying)

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    5 g

Specifications