Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bromphenol blå natriumsalt

Bromphenol blå natriumsalt

Glass bottle, 1 g
Katalog # TCIAS0043-1G
Leverandør:   TCI
Bromphenol blå natriumsalt
Katalog # TCIAS0043-1G
Leverandør:  TCI
CAS nummer:  34725-61-6
Bromphenol blå natriumsalt
Bromphenol blå natriumsalt

Glass bottle, 1 g

CAS nummer:  34725-61-6
Price
255,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents
Synonymer:  Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt
  • CAS nummer:34725-61-6
  • Formel:C₁₉H₉Br₄NaO₅S
  • MDL nummer:MFCD00013793
  • Molecular Weight:691.95 g/mol
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:252-170-2
Specifikationer

Specifikationstestresultater

  • Appearance:
    Dark green to Dark blue to Black powder to crystal
  • Absorbance:
    min. 0.480 (5 mg/L, pH 5.0, 590.0 to 594.0 nm)
  • Solubility in Water:
    almost transparency
  • Drying loss:
    max. 7.0 %
  • Ignition residue (Sulfate):
    10.0 to 10.8 % (after drying)

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.