Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Saponin

Glass bottle, 250 g
  TCIAS0019-250G
 :   TCI
Saponin
  TCIAS0019-250G
 :  TCI
 :  8047-15-2
Saponin

Glass bottle, 250 g

 :  8047-15-2
 
3.630,00 kr
   
1 * 250 g
 
  •  :8047-15-2
  •  :MFCD00081981
  •  :1,015 to 1.02 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :232-462-6
 

 

  • Drying loss:
    max. 10.0 %
  • Ignition residue (Sulfate):
    max. 20.0 %
  • Sapogenine:
    min. 9.0 %

 

  • Pakning:
    Glass bottle
  • Pakn:
    250 g

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.