Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Saponin

Glass bottle, 25 g
Katalog # TCIAS0019-25G
Leverandør:   TCI
Saponin
Katalog # TCIAS0019-25G
Leverandør:  TCI
CAS nummer:  8047-15-2
Saponin

Glass bottle, 25 g

CAS nummer:  8047-15-2
Price
460,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
  • CAS nummer:8047-15-2
  • MDL nummer:MFCD00081981
  • Tæthed:1,015 to 1.02 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:232-462-6
Specifikationer

Specifikationstestresultater

  • Drying loss:
    max. 10.0 %
  • Ignition residue (Sulfate):
    max. 20.0 %
  • Sapogenine:
    min. 9.0 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.