Phenol rød natrium salt
Glass bottle, 5 g
Katalog # TCIAP0102-5G
Leverandør: TCI
Gå til:
Product Details & Documents
Synonymer: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide
- CAS nummer:34487-61-1
- Formel:C₁₉H₁₃NaO₅S
- MDL nummer:MFCD00066901
- Molecular Weight:376.35 g/mol
- Merck Index:12,07397
- Smeltende Pt:285 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:252-057-8
Specifikationer
Specifikationstestresultater
- Appearance:Orange to Amber to Dark red powder to crystal
- Absorbance:min. 0.530 (5 mg/L, pH8.4, 556.0 to 560.0 nm)
- Solubility in Water:almost transparency
- Drying loss:max. 5.0 %
- Transition interval (pH):(redish yellow)6.8 to 8.4 (red)
Specifikationer
- Pakning:Glass bottle
- Pakn:5 g



