Phenol rød natrium salt
Glass bottle, 25 g
Katalog # TCIAP0102-25G
Leverandør: TCI
Gå til:
Product Details & Documents
Synonymer: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide
- CAS nummer:34487-61-1
- Formel:C₁₉H₁₃NaO₅S
- MDL nummer:MFCD00066901
- Molecular Weight:376.35 g/mol
- Merck Index:12,07397
- Smeltende Pt:285 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:252-057-8
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Orange to Amber to Dark red powder to crystal
- Absorbance:min. 0.530 (5 mg/L, pH8.4, 556.0 to 560.0 nm)
- Solubility in Water:almost transparency
- Drying loss:max. 5.0 %
- Transition interval (pH):(redish yellow)6.8 to 8.4 (red)
Specifikationer
- Pakning:Glass bottle
- Pakn:25 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



