Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumpyruvat ≥97.0% (by titrimetric analysis)

Natriumpyruvat ≥97.0% (by titrimetric analysis)

Glass bottle, 500 g
Catalog # TCIAP0582-500G
Supplier:   TCI
Natriumpyruvat ≥97.0% (by titrimetric analysis)
Catalog # TCIAP0582-500G
Supplier:  TCI
CAS Number:  113-24-6
Natriumpyruvat ≥97.0% (by titrimetric analysis)
Natriumpyruvat ≥97.0% (by titrimetric analysis)

Glass bottle, 500 g

CAS Number:  113-24-6
Price
977,00 kr
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
  • CAS Number:113-24-6
  • Formula:CH₃COCOONa
  • MDL Number:MFCD00002586
  • Molecular Weight:110.04 g/mol
  • Melting Pt:220…230 °C
  • Density:1,718 g/cm³ (20 °C)
  • Storage Temperature:Køleskab
  • EINECS:204-024-4
Specifications

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (Iodometric Titration):
    min. 97.0 %
  • Solubility in Water:
    almost transparency

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    500 g

Specifications