Natriumpyruvat ≥97.0% (by titrimetric analysis)
Glass bottle, 25 g
TCIAP0582-25G
: TCI
: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- :113-24-6
- :CH₃COCOONa
- :MFCD00002586
- :110.04 g/mol
- :220…230 °C
- :1,718 g/cm³ (20 °C)
- :Køleskab
- :204-024-4
- Appearance:White powder to crystal
- Purity (Iodometric Titration):min. 97.0 %
- Solubility in Water:almost transparency
- Pakning:Glass bottle
- Pakn:25 g



