Nile red
500 mg
Katalog # TCIAN0659-500MG
Leverandør: TCI
Gå til:
Product Details & Documents
Synonymer: 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one
- CAS nummer:7385-67-3
- Formel:C₂₀H₁₈N₂O₂
- MDL nummer:MFCD00011639
- Molecular Weight:318.38 g/mol
- EINECS:230-966-0
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Yellow to Brown to Dark green powder to crystal
- Lambda max.:550.0 to 554.0 nm (MeOH)
- Absorbance (E1%1cm):min. 1,300 (MeOH, 550.0 to 554.0 nm)
Specifikationer
- Pakn:500 mg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



