Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Pentadecylcyclohexan ≥98.0% (by GC)

Glass bottle, 5 g
Katalog # TCIAP0369-5G
Leverandør:   TCI
Pentadecylcyclohexan ≥98.0% (by GC)
Katalog # TCIAP0369-5G
Leverandør:  TCI
CAS nummer:  6006-95-7
Pentadecylcyclohexan ≥98.0% (by GC)
Pentadecylcyclohexan ≥98.0% (by GC)

Glass bottle, 5 g

CAS nummer:  6006-95-7
Price
1.520,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
  • CAS nummer:6006-95-7
  • Formel:C₂₁H₄₂
  • MDL nummer:MFCD00058952
  • Molecular Weight:294.56 g/mol
  • Kogende Pt:373 °C (1013 hPa)
  • Smeltende Pt:29 °C
  • Tæthed:1,03 g/cm³ (0 °C)
  • EINECS:227-858-0
Specifikationer

Specifikationstestresultater

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %
  • Freezing point:
    24.0 to 26.0 deg-C

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    5 g

Specifikationer