Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Pentadecylcyclohexan ≥98.0% (by GC)

Pentadecylcyclohexan ≥98.0% (by GC)

Glass bottle, 25 g
Catalog # TCIAP0369-25G
Supplier:   TCI
Pentadecylcyclohexan ≥98.0% (by GC)
Catalog # TCIAP0369-25G
Supplier:  TCI
CAS Number:  6006-95-7
Pentadecylcyclohexan ≥98.0% (by GC)
Pentadecylcyclohexan ≥98.0% (by GC)

Glass bottle, 25 g

CAS Number:  6006-95-7
Price
5.910,00 kr
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:6006-95-7
  • Formula:C₂₁H₄₂
  • MDL Number:MFCD00058952
  • Molecular Weight:294.56 g/mol
  • Boiling Pt:373 °C (1013 hPa)
  • Melting Pt:29 °C
  • Density:1,03 g/cm³ (0 °C)
  • EINECS:227-858-0
Specifications

Specification Test Results

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %
  • Freezing point:
    24.0 to 26.0 deg-C

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.