Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

4-Methylmorpholin ≥99.0% (by GC)

Glass bottle, 25 mL
Katalog # TCIAM0370-25ML
Brandfarlig
Ætsende
Irriterende
Leverandør:   TCI
4-Methylmorpholin ≥99.0% (by GC)
Katalog # TCIAM0370-25ML
Leverandør:  TCI
CAS nummer:  109-02-4
4-Methylmorpholin ≥99.0% (by GC)
4-Methylmorpholin ≥99.0% (by GC)

Glass bottle, 25 mL

CAS nummer:  109-02-4
Price
140,00 kr
Log ind for tilgængelighed
1 * 25 mL
Product Details & Documents
Synonymer:  NMM, N-Methylmorpholine
  • CAS nummer:109-02-4
  • Formel:C₅H₁₁NO
  • MDL nummer:MFCD00006175
  • Molecular Weight:101.15 g/mol
  • Merck Index:14,06277
  • Kogende Pt:116°C
  • Smeltende Pt:–66 °C
  • Tæthed:0,922 g/cm³ (20 °C)
  • Flash Pt:18°C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-640-0
  • FN:2535
  • ADR:3,II
Specifikationer

Specifikationstestresultater

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 99.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    25 mL

Specifikationer