Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Methylcyclohexan ≥99.0% (by GC)

Glass bottle, 500 mL
Katalog # TCIAM0190-500ML
Brandfarlig
Irriterende
Sundhed
Miljø
Leverandør:   TCI
Methylcyclohexan ≥99.0% (by GC)
Katalog # TCIAM0190-500ML
Leverandør:  TCI
CAS nummer:  108-87-2
Methylcyclohexan ≥99.0% (by GC)
Methylcyclohexan ≥99.0% (by GC)

Glass bottle, 500 mL

CAS nummer:  108-87-2
Price
329,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
  • CAS nummer:108-87-2
  • Formel:C₆H₁₁CH₃
  • MDL nummer:MFCD00001497
  • Molecular Weight:98.19 g/mol
  • Merck Index:14,06047
  • Kogende Pt:100°C
  • Smeltende Pt:–127°C
  • Smeltende Pt:-127°C
  • Tæthed:0,7693 g/cm³ (20 °C)
  • Flash Pt:–4°C
  • Flash Pt:-4°C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-624-3
  • FN:2296
  • ADR:3,II
Specifikationer

Specifikationstestresultater

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.0 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.