Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Methylcyclohexan ≥99.0% (by GC)

Methylcyclohexan ≥99.0% (by GC)

Glass bottle, 25 mL
Katalog # TCIAM0190-25ML
Brandfarlig
Irriterende
Sundhed
Miljø
Leverandør:   TCI
Methylcyclohexan ≥99.0% (by GC)
Katalog # TCIAM0190-25ML
Leverandør:  TCI
CAS nummer:  108-87-2
Methylcyclohexan ≥99.0% (by GC)
Methylcyclohexan ≥99.0% (by GC)

Glass bottle, 25 mL

CAS nummer:  108-87-2
Price
kr132.00
Log ind for tilgængelighed
1 * 25 mL
Product Details & Documents
  • CAS nummer:108-87-2
  • Formel:C₆H₁₁CH₃
  • MDL nummer:MFCD00001497
  • Molecular Weight:98.19 g/mol
  • Merck Index:14,06047
  • Kogende Pt:100°C
  • Smeltende Pt:–127°C
  • Smeltende Pt:-127°C
  • Tæthed:0,7693 g/cm³ (20 °C)
  • Flash Pt:–4°C
  • Flash Pt:-4°C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-624-3
  • FN:2296
  • ADR:3,II
Specifikationer

Specifikationstestresultater

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.0 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    25 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.