Metanil gul
Glass bottle, 500 g
TCIAM0490-500G
: TCI
: Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
- :587-98-4
- :C₁₈H₁₄N₃NaO₃S
- :MFCD00007486
- :375.38 g/mol
- :14,05928
- :209-608-2
Documents
- Appearance:Light yellow to Brown powder to crystal
- Lambda max.:415.0 to 419.0 nm (MeOH)
- Absorbance (E1%1cm):min. 500 (MeOH, 415.0 to 419.0 nm)
- Pakning:Glass bottle
- Pakn:500 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



