Metanil gul
Glass bottle, 25 g
Katalog # TCIAM0490-25G
Leverandør: TCI
Gå til:
Product Details & Documents
Synonymer: Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
- CAS nummer:587-98-4
- Formel:C₁₈H₁₄N₃NaO₃S
- MDL nummer:MFCD00007486
- Molecular Weight:375.38 g/mol
- Merck Index:14,05928
- EINECS:209-608-2
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Light yellow to Brown powder to crystal
- Lambda max.:415.0 to 419.0 nm (MeOH)
- Absorbance (E1%1cm):min. 500 (MeOH, 415.0 to 419.0 nm)
Specifikationer
- Pakning:Glass bottle
- Pakn:25 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



