Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Morpholin ≥99.0% (by GC, titration analysis)

Glass bottle, 500 mL
Katalog # TCIAM0465-500ML
Brandfarlig
Ætsende
Giftig
Sundhed
Leverandør:   TCI
Morpholin ≥99.0% (by GC, titration analysis)
Katalog # TCIAM0465-500ML
Leverandør:  TCI
CAS nummer:  110-91-8
Morpholin ≥99.0% (by GC, titration analysis)
Morpholin ≥99.0% (by GC, titration analysis)

Glass bottle, 500 mL

CAS nummer:  110-91-8
Price
197,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
  • CAS nummer:110-91-8
  • Formel:C₄H₉NO
  • MDL nummer:MFCD00005972
  • Molecular Weight:87.12 g/mol
  • Merck Index:14,06277
  • Kogende Pt:129°C
  • Smeltende Pt:–5 °C
  • Tæthed:1 g/cm³ (20 °C)
  • Flash Pt:32°C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-815-1
  • FN:2054
  • ADR:8,I
Specifikationer

Specifikationstestresultater

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %
  • Purity (Neutralization titration):
    min. 99.0 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.