Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Morpholin ≥99.0% (by GC, titration analysis)

Morpholin ≥99.0% (by GC, titration analysis)

Glass bottle, 25 mL
Katalog # TCIAM0465-25ML
Brandfarlig
Ætsende
Giftig
Sundhed
Leverandør:   TCI
Morpholin ≥99.0% (by GC, titration analysis)
Katalog # TCIAM0465-25ML
Leverandør:  TCI
CAS nummer:  110-91-8
Morpholin ≥99.0% (by GC, titration analysis)
Morpholin ≥99.0% (by GC, titration analysis)

Glass bottle, 25 mL

CAS nummer:  110-91-8
Price
132,00 kr
Log ind for tilgængelighed
1 * 25 mL
Product Details & Documents
  • CAS nummer:110-91-8
  • Formel:C₄H₉NO
  • MDL nummer:MFCD00005972
  • Molecular Weight:87.12 g/mol
  • Merck Index:14,06277
  • Kogende Pt:129°C
  • Smeltende Pt:–5 °C
  • Tæthed:1 g/cm³ (20 °C)
  • Flash Pt:32°C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-815-1
  • FN:2054
  • ADR:8,I
Specifikationer

Specifikationstestresultater

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %
  • Purity (Neutralization titration):
    min. 99.0 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    25 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.