Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(±)-Linalool ≥96.0% (by GC)

(±)-Linalool ≥96.0% (by GC)

Glass bottle, 500 mL
Katalog # TCIAL0048-500ML
Irriterende
Sundhed
Leverandør:   TCI
(±)-Linalool ≥96.0% (by GC)
Katalog # TCIAL0048-500ML
Leverandør:  TCI
CAS nummer:  78-70-6
(±)-Linalool ≥96.0% (by GC)
(±)-Linalool ≥96.0% (by GC)

Glass bottle, 500 mL

CAS nummer:  78-70-6
Price
690,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
Synonymer:  (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol
  • CAS nummer:78-70-6
  • Formel:C₁₀H₁₈O
  • MDL nummer:MFCD00008906
  • Molecular Weight:154.25 g/mol
  • Merck Index:14,05495
  • Kogende Pt:200°C
  • Tæthed:0,87 g/cm³ (20 °C)
  • Flash Pt:71°C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-134-4
Specifikationer

Specifikationstestresultater

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 96.0 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.