Lecithin (from soy bean)
Glass bottle, 500 g
Katalog # TCIAL0023-500G
Leverandør: TCI
Gå til:
Product Details & Documents
- CAS nummer:8002-43-5
- MDL nummer:MFCD00147406
- Merck Index:14,05429
- EINECS:232-307-2
Specifikationer
Specifikationstestresultater
- Appearance:Yellow to Amber to Dark purple clear liquid to slightly cloudy liquid
- Saponification Value:180.0 to 195.0
- Acetone insoluble matter:58.0 to 65.0 %
- Acid Value:max. 25.0
Specifikationer
- Pakning:Glass bottle
- Pakn:500 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



