Lecithin (from soy bean)
Glass bottle, 25 g
: TCI
- :8002-43-5
- :MFCD00147406
- :14,05429
- :232-307-2
- Appearance:Yellow to Amber to Dark purple clear liquid to slightly cloudy liquid
- Saponification Value:180.0 to 195.0
- Acetone insoluble matter:58.0 to 65.0 %
- Acid Value:max. 25.0
- Pakning:Glass bottle
- Pakn:25 g



