Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Finasteride ≥98.0% (by GC)

Finasteride ≥98.0% (by GC)

200 mg
Katalog # TCIAF0675-200MG
Irriterende
Leverandør:   TCI
Finasteride ≥98.0% (by GC)
Katalog # TCIAF0675-200MG
Leverandør:  TCI
CAS nummer:  98319-26-7
Finasteride ≥98.0% (by GC)

200 mg

CAS nummer:  98319-26-7
Price
255,00 kr
Log ind for tilgængelighed
1 * 200 mg
Product Details & Documents
Synonymer:  N-tert-Butyl-3-oxo-4-aza-5alpha-androst-1-en-17beta-carboxamide
  • CAS nummer:98319-26-7
  • Formel:C₂₃H₃₆N₂O₂
  • MDL nummer:MFCD00869737
  • Molecular Weight:372.55 g/mol
  • Merck Index:14,04082
  • Smeltende Pt:253°C
Specifikationer

Specifikationstestresultater

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %

Specifikationer

  • Pakn:
    200 mg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.