Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Finasteride ≥98.0% (by GC)

Finasteride ≥98.0% (by GC)

1 g
Katalog # TCIAF0675-1G
Irriterende
Leverandør:   TCI
Finasteride ≥98.0% (by GC)
Katalog # TCIAF0675-1G
Leverandør:  TCI
CAS nummer:  98319-26-7
Finasteride ≥98.0% (by GC)

1 g

CAS nummer:  98319-26-7
Price
952,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents
Synonymer:  N-tert-Butyl-3-oxo-4-aza-5alpha-androst-1-en-17beta-carboxamide
  • CAS nummer:98319-26-7
  • Formel:C₂₃H₃₆N₂O₂
  • MDL nummer:MFCD00869737
  • Molecular Weight:372.55 g/mol
  • Merck Index:14,04082
  • Smeltende Pt:253°C
Specifikationer

Specifikationstestresultater

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %

Specifikationer

  • Pakn:
    1 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.