Ethidiumbromid ≥95.0% (by HPLC, titration analysis)
Glass bottle, 5 g
Katalog # TCIAE0370-5G
Leverandør: TCI
Ethidiumbromid ≥95.0% (by HPLC, titration analysis)
Katalog # TCIAE0370-5G
Leverandør: TCI
CAS nummer: 1239-45-8
Gå til:
Product Details & Documents
Synonymer: 3,8-Diamino-5-ethyl-6-phenylphenantridiniumbromid
- CAS nummer:1239-45-8
- Formel:C₂₁H₂₀BrN₃
- MDL nummer:MFCD00011724
- Molecular Weight:394.31 g/mol
- Merck Index:14,04731
- Kogende Pt:72 °C (0,1 -rr)
- Smeltende Pt:261…264 °C
- Tæthed:1,01 g/cm³ (20 °C)
- Flash Pt:≤ 100 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:214-984-6
- FN:1544
- ADR:6.1,I
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Orange to Brown to Dark purple powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Argentmetric Titration):min. 95.0 %
Specifikationer
- Pakning:Glass bottle
- Pakn:5 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



