Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Ethidiumbromid ≥95.0% (by HPLC, titration analysis)

Glass bottle, 1 g
Katalog # TCIAE0370-1G
Giftig
Sundhed
Leverandør:   TCI
Ethidiumbromid ≥95.0% (by HPLC, titration analysis)
Katalog # TCIAE0370-1G
Leverandør:  TCI
CAS nummer:  1239-45-8
Ethidiumbromid ≥95.0% (by HPLC, titration analysis)
Ethidiumbromid ≥95.0% (by HPLC, titration analysis)

Glass bottle, 1 g

CAS nummer:  1239-45-8
Price
673,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents
Synonymer:  3,8-Diamino-5-ethyl-6-phenylphenantridiniumbromid
  • CAS nummer:1239-45-8
  • Formel:C₂₁H₂₀BrN₃
  • MDL nummer:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Merck Index:14,04731
  • Kogende Pt:72 °C (0,1 -rr)
  • Smeltende Pt:261…264 °C
  • Tæthed:1,01 g/cm³ (20 °C)
  • Flash Pt:≤ 100 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:214-984-6
  • FN:1544
  • ADR:6.1,I
Specifikationer

Specifikationstestresultater

  • Appearance:
    Orange to Brown to Dark purple powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Purity (Argentmetric Titration):
    min. 95.0 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 g

Specifikationer