Glutaraldehyd 24 - 26% in water
Glass bottle, 500 mL
Katalog # TCIAG0067-500ML
Leverandør: TCI
Gå til:
Product Details & Documents
Synonymer: Glutaral
- CAS nummer:111-30-8
- Formel:C₅H₈O₂
- MDL nummer:MFCD00007025
- Molecular Weight:100.12 g/mol
- Merck Index:14,04472
- Kogende Pt:101 °C (1013 hPa)
- Smeltende Pt:–6 °C
- Tæthed:1,06 g/cm³ (20 °C)
- Flash Pt:≥ 71 °C
- Opbevaringstemperatur:Køleskab
- FN:2927
- ADR:6.1,II
- :Density: 1.014 g/cm³ (20 °C)
- EINECS:203-856-5
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Colorless clear liquid
- Concentration (Oxime Formation):24.0 to 26.0 w/w%
- abs-ratio (a/b):max. 0.35 (E230/280)
Specifikationer
- Pakning:Glass bottle
- Pakn:500 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



