Digoxin ≥96.0% (by HPLC)
Glass bottle, 100 mg
Katalog # TCIAD1828-100MG
Leverandør: TCI
Gå til:
Product Details & Documents
- CAS nummer:20830-75-5
- Formel:C₄₁H₆₄O₁₄
- MDL nummer:MFCD00003674
- Molecular Weight:780.95 g/mol
- Merck Index:14,03167
- Smeltende Pt:265°C
- EINECS:244-068-1
- FN:1544
- ADR:6.1,II
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White to Almost white powder to crystaline
- Purity (HPLC):min. 96.0 area%
- Specific rotation [a]20/D:+10.0 to +13.0 deg (C=10, Pyridine)
- Solubility in Pyridine:almost transparency
Specifikationer
- Pakning:Glass bottle
- Pakn:100 mg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



