Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Cyclohexan ≥99.5% (by GC)

Glass bottle, 25 mL
  TCIAC0469-25ML
 
 
 
 
 :   TCI
Cyclohexan ≥99.5% (by GC)
  TCIAC0469-25ML
 :  TCI
 :  110-82-7
Cyclohexan ≥99.5% (by GC)
Cyclohexan ≥99.5% (by GC)

Glass bottle, 25 mL

 :  110-82-7
 
132,00 kr
   
1 * 25 mL
 
 :  Hexahydrobenzene, Hexamethylene
  •  :110-82-7
  •  :C₆H₁₂
  •  :MFCD00003814
  •  :84.16 g/mol
  •  :14,02723
  •  :81°C
  •  :7 °C
  •  :0,7785 g/cm³ (20 °C)
  •  :–20°C
  •  :-20°C
  •  :Stue temperatur
  •  :203-806-2
  •  :1145
  •  :3,II
 

 

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.5 %

 

  • Pakning:
    Glass bottle
  • Pakn:
    25 mL

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.