Basic Violet 3
25 g
Katalog # TCIAC0428-25G
Leverandør: TCI
Gå til:
Product Details & Documents
Synonymer: 4-[4,4'-Bis(dimethylamino)benzhydryliden]cyclohexa-2,5-dien-1-yliden]dimethylammoniumchlorid, CI 42555
- CAS nummer:548-62-9
- Formel:C₂₅H₃₀ClN₃
- MDL nummer:MFCD00011750
- Molecular Weight:407.99 g/mol
- Merck Index:14,04395
- Smeltende Pt:211°C
- EINECS:208-953-6
- FN:3077
- ADR:9,III
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Green to Dark green powder to crystal
- Absorbance:min. 0.300 (2mg/L, 588.0 to 592.0 nm) (calcd.on dried substance)
- Ethanol insoluble matter:max. 20.0 %
- Drying loss:max. 10.0 %
- Sensitiveness:Turn Purple in Acetic Acid and turn Yellow green by adding Perchloric Acid
Specifikationer
- Pakn:25 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



