Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2-Chlorhexan ≥98.0% (by GC)

2-Chlorhexan ≥98.0% (by GC)

Glass bottle, 5 g
Catalog # TCIAC0890-5G
Flammable
Irritant
Supplier:   TCI
2-Chlorhexan ≥98.0% (by GC)
Catalog # TCIAC0890-5G
Supplier:  TCI
CAS Number:  638-28-8
2-Chlorhexan ≥98.0% (by GC)
2-Chlorhexan ≥98.0% (by GC)

Glass bottle, 5 g

CAS Number:  638-28-8
Price
788,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2-Hexyl chloride
  • CAS Number:638-28-8
  • Formula:C₆H₁₃Cl
  • MDL Number:MFCD00039351
  • Molecular Weight:120.62 g/mol
  • Boiling Pt:122°C
  • Storage Temperature:Køleskab
  • EINECS:211-329-6
  • UN:1993
  • ADR:3,III
Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    5 g

Specifications