Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

2-Chlorhexan ≥98.0% (by GC)

Glass bottle, 25 g
  TCIAC0890-25G
 
 
 :   TCI
2-Chlorhexan ≥98.0% (by GC)
  TCIAC0890-25G
 :  TCI
 :  638-28-8
2-Chlorhexan ≥98.0% (by GC)
2-Chlorhexan ≥98.0% (by GC)

Glass bottle, 25 g

 :  638-28-8
 
2.580,00 kr
   
1 * 25 g
 
 :  2-Hexyl chloride
  •  :638-28-8
  •  :C₆H₁₃Cl
  •  :MFCD00039351
  •  :120.62 g/mol
  •  :122°C
  •  :Køleskab
  •  :211-329-6
  •  :1993
  •  :3,III
 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

  • Pakning:
    Glass bottle
  • Pakn:
    25 g