Chlorophenol rød ≥93.0% (by HPLC)
Glass bottle, 5 g
Katalog # TCIAC0245-5G
Leverandør: TCI
Gå til:
Product Details & Documents
Synonymer: 3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
- CAS nummer:4430-20-0
- Formel:C₁₉H₁₂Cl₂O₅S
- MDL nummer:MFCD00005877
- Molecular Weight:423.27 g/mol
- Smeltende Pt:∼261 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:224-619-2
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 93.0 area%
- Absorbance:min. 0.520 (4 mg/L, pH7.0, 573.0 to 577.0 nm)
- Transition interval (pH):(yellow)5.0 to 6.6 (red purple)
Specifikationer
- Pakning:Glass bottle
- Pakn:5 g



