Chlorophenol rød ≥93.0% (by HPLC)
Glass bottle, 1 g
Katalog # TCIAC0245-1G
Leverandør: TCI
Gå til:
Product Details & Documents
Synonymer: 3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
- CAS nummer:4430-20-0
- Formel:C₁₉H₁₂Cl₂O₅S
- MDL nummer:MFCD00005877
- Molecular Weight:423.27 g/mol
- Smeltende Pt:∼261 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:224-619-2
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 93.0 area%
- Absorbance:min. 0.520 (4 mg/L, pH7.0, 573.0 to 577.0 nm)
- Transition interval (pH):(yellow)5.0 to 6.6 (red purple)
Specifikationer
- Pakning:Glass bottle
- Pakn:1 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



