Bromphenol blå
Glass bottle, 25 g
Catalog # TCIAB0631-25G
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
- CAS Number:115-39-9
- Formula:C₁₉H₁₀Br₄O₅S
- MDL Number:MFCD00005875
- Molecular Weight:669.97 g/mol
- Merck Index:14,01444
- Melting Pt:273 °C
- Density:1 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:204-086-2
Documents
Specifications
Specification Test Results
- Appearance:Light red to Yellow to Red powder to crystal
- Absorbance:min. 0.480 (5 mg/L, pH 5.0, 590.0 to 594.0 nm)
- Solubility in Sodium hydroxide solution:transparency
- Solubility in EtOH (50vol%):transparency
- Drying loss:max. 1.0 %
Specifications
- Pakning:Glass bottle
- Pakn:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



