Bromcresol grønt
Glass bottle, 25 g
: TCI
: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- :76-60-8
- :C₂₁H₁₄Br₄O₅S
- :MFCD00005874
- :698.02 g/mol
- :14,01386
- :217…218 °C
- :Stue temperatur
- :200-972-8
- Appearance:White to Gray to Brown powder to crystal
- Absorbance:min. 0.250 (5mg/L, pH5.8, 613.0 to 617.0 nm)
- Solubility in Sodium hydroxide solution:almost transparency
- Drying loss:max. 1.0 %
- Ignition residue (Sulfate):max. 0.5 %
- Pakning:Glass bottle
- Pakn:25 g



