Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bromcresol grønt

Bromcresol grønt

Glass bottle, 1 g
Katalog # TCIAB0578-1G
Leverandør:   TCI
Bromcresol grønt
Katalog # TCIAB0578-1G
Leverandør:  TCI
CAS nummer:  76-60-8
Bromcresol grønt
Bromcresol grønt

Glass bottle, 1 g

CAS nummer:  76-60-8
Price
132,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents
Synonymer:  3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS nummer:76-60-8
  • Formel:C₂₁H₁₄Br₄O₅S
  • MDL nummer:MFCD00005874
  • Molecular Weight:698.02 g/mol
  • Merck Index:14,01386
  • Smeltende Pt:217…218 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-972-8
Specifikationer

Specifikationstestresultater

  • Appearance:
    White to Gray to Brown powder to crystal
  • Absorbance:
    min. 0.250 (5mg/L, pH5.8, 613.0 to 617.0 nm)
  • Solubility in Sodium hydroxide solution:
    almost transparency
  • Drying loss:
    max. 1.0 %
  • Ignition residue (Sulfate):
    max. 0.5 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.