L (+)-Ascorbinsyre ≥99.0% (by titrimetric analysis)
Glass bottle, 500 g
: TCI
: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- :50-81-7
- :C₆H₈O₆
- :MFCD00064328
- :176.13 g/mol
- :14,00830
- :553 °C (1013 hPa)
- :192°C
- :1,694 g/cm³ (20 °C)
- :Stue temperatur
- :200-066-2
- :exempt
- Appearance:White powder to crystal
- Purity (Iodometric Titration):min. 99.0 %
- Specific rotation [a]20/D:+20.5 to +21.5deg (C=10, H2O)
- Solubility in Water:almost transparency
- Pakning:Glass bottle
- Pakn:500 g



