L (+)-Ascorbinsyre ≥99.0% (by titrimetric analysis)
Glass bottle, 25 g
Katalog # TCIAA0537-25G
Leverandør: TCI
L (+)-Ascorbinsyre ≥99.0% (by titrimetric analysis)
Katalog # TCIAA0537-25G
Leverandør: TCI
CAS nummer: 50-81-7
Gå til:
Product Details & Documents
Synonymer: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS nummer:50-81-7
- Formel:C₆H₈O₆
- MDL nummer:MFCD00064328
- Molecular Weight:176.13 g/mol
- Merck Index:14,00830
- Kogende Pt:553 °C (1013 hPa)
- Smeltende Pt:192°C
- Tæthed:1,694 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-066-2
- NÅ:exempt
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White powder to crystal
- Purity (Iodometric Titration):min. 99.0 %
- Specific rotation [a]20/D:+20.5 to +21.5deg (C=10, H2O)
- Solubility in Water:almost transparency
Specifikationer
- Pakning:Glass bottle
- Pakn:25 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



