Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Anthron ≥98.0% (by GC)

Anthron ≥98.0% (by GC)

Glass bottle, 25 g
Katalog # TCIAA0509-25G
Irriterende
Leverandør:   TCI
Anthron ≥98.0% (by GC)
Katalog # TCIAA0509-25G
Leverandør:  TCI
CAS nummer:  90-44-8
Anthron ≥98.0% (by GC)
Anthron ≥98.0% (by GC)

Glass bottle, 25 g

CAS nummer:  90-44-8
Price
1.360,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  Anthracen-9(10H)-one
  • CAS nummer:90-44-8
  • Formel:C₁₄H₁₀O
  • MDL nummer:MFCD00001187
  • Molecular Weight:194.23 g/mol
  • Merck Index:14,00691
  • EINECS:201-994-0

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    154.0 to 160.0 deg-C
  • Solubility in hot Toluene:
    almost transparency
  • suitability for glucose-analysis:
    Abs min. 0.70 (625nm ) in the presence of D-Glucose (10 ppm).

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    25 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.